Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332892.1 | NP_001319111.1 | 24 - 25 | 4214 | 102 | |
NM_001332893.1 | NP_001323123.1 | 24 - 25 | 4327 | 102 | ||
NM_001332894.1 | NP_001323127.1 | 24 - 25 | 4311 | 102 | ||
NM_001332895.1 | NP_001323124.1 | 24 - 25 | 4311 | 102 | ||
NM_001332896.1 | NP_001323125.1 | 23 - 24 | 4431 | 102 | ||
NM_001332897.1 | NP_001323126.1 | 23 - 24 | 3966 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |