Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332892.1 | NP_001319111.1 | 5 - 6 | 950 | 109 | |
NM_001332893.1 | NP_001323123.1 | 5 - 6 | 1072 | 109 | ||
NM_001332894.1 | NP_001323127.1 | 5 - 6 | 1056 | 109 | ||
NM_001332895.1 | NP_001323124.1 | 5 - 6 | 1056 | 109 | ||
NM_001332896.1 | NP_001323125.1 | 4 - 5 | 1176 | 109 | ||
NM_001332897.1 | NP_001323126.1 | 4 - 5 | 711 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |