Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332900.1 | NP_001323337.1 | 8 - 9 | 1181 | 63 | |
NM_102781.3 | NP_174333.2 | 10 - 11 | 1481 | 63 | ||
NM_179400.1 | NP_849731.1 | 10 - 11 | 1450 | 63 | ||
NM_179401.2 | NP_849732.1 | 10 - 11 | 1481 | 63 | ||
GenBank mRNA | AK226602.1 | - | 10 - 11 | 1436 | 63 | |
AM113986.1 | - | 10 - 11 | 1451 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |