Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102785.5 | NP_174337.1 | 3 - 4 | 842 | 74 | |
GenBank mRNA | AJ440967.1 | - | 3 - 4 | 493 | 74 | |
AK226570.1 | - | 3 - 4 | 707 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |