Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102820.3 | NP_564364.1 | 3 - 4 | 528 | 142 | |
GenBank mRNA | AK118475.1 | - | 3 - 4 | 492 | 142 | |
AY084256.1 | - | 3 - 4 | 492 | 142 | ||
BT005308.1 | - | 3 - 4 | 330 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |