Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084165.1 | NP_001077634.1 | 4 - 5 | 982 | 67 | |
NM_001332933.1 | NP_001322001.1 | 4 - 5 | 1022 | 67 | ||
NM_102836.3 | NP_564369.1 | 4 - 5 | 1000 | 67 | ||
GenBank mRNA | AF360277.1 | - | 4 - 5 | 998 | 67 | |
AY051039.1 | - | 4 - 5 | 842 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |