Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084168.1 | NP_001077637.1 | 2 - 3 | 272 | 126 | |
NM_001198192.1 | NP_001185121.1 | 2 - 3 | 347 | 126 | ||
NM_001332946.1 | NP_001321339.1 | 2 - 3 | 272 | 126 | ||
NM_102855.4 | NP_564375.1 | 2 - 3 | 297 | 126 | ||
GenBank mRNA | AK117789.1 | - | 2 - 3 | 243 | 126 | |
BT004731.1 | - | 2 - 3 | 236 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |