Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102887.5 | NP_174433.2 | 14 - 15 | 2245 | 86 | |
GenBank mRNA | AB073133.1 | - | 14 - 15 | 2095 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |