Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102887.5 | NP_174433.2 | 21 - 22 | 2771 | 102 | |
GenBank mRNA | AB073133.1 | - | 21 - 22 | 2621 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |