Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP97A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102914.3 | NP_564384.1 | 2 - 3 | 487 | 117 | |
GenBank mRNA | AY056446.1 | - | 2 - 3 | 482 | 117 | |
AY058173.1 | - | 2 - 3 | 487 | 117 | ||
AY142017.1 | - | 2 - 3 | 449 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |