Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP97A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102914.3 | NP_564384.1 | 7 - 8 | 1125 | 89 | |
GenBank mRNA | AY056446.1 | - | 7 - 8 | 1120 | 89 | |
AY058173.1 | - | 7 - 8 | 1125 | 89 | ||
AY142017.1 | - | 7 - 8 | 1087 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |