Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102916.4 | NP_174462.1 | 2 - 3 | 263 | 75 | |
GenBank mRNA | AF326905.1 | - | 2 - 3 | 185 | 75 | |
AF339724.1 | - | 1 - 2 | 129 | 75 | ||
AF380634.1 | - | 2 - 3 | 185 | 75 | ||
AY054132.1 | - | 1 - 2 | 129 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |