Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333002.1 | NP_001322279.1 | 4 - 5 | 353 | 132 | |
NM_102957.4 | NP_564392.1 | 5 - 6 | 1429 | 132 | ||
GenBank mRNA | AB493491.1 | - | 5 - 6 | 1039 | 132 | |
AY048689.1 | - | 5 - 6 | 1057 | 132 | ||
BT029211.1 | - | 5 - 6 | 1039 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Arabidopsis thaliana Probe spans exons |