Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198198.1 | NP_001185127.1 | 5 - 6 | 713 | 92 | |
NM_102984.5 | NP_174527.2 | 5 - 6 | 708 | 92 | ||
GenBank mRNA | AY074318.1 | - | 5 - 6 | 631 | 92 | |
AY133872.1 | - | 4 - 5 | 485 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |