Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sks9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342496.1 | NP_001329209.1 | 4 - 5 | 752 | 82 | |
NM_001342497.1 | NP_001329210.1 | 4 - 5 | 694 | 82 | ||
NM_120004.5 | NP_195555.2 | 4 - 5 | 852 | 82 | ||
GenBank mRNA | AY072215.1 | - | 4 - 5 | 694 | 82 | |
AY096593.1 | - | 4 - 5 | 628 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |