Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GH9B17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120060.2 | NP_195611.1 | 4 - 5 | 1196 | 86 | |
GenBank mRNA | DQ446906.1 | - | 4 - 5 | 1153 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |