Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120063.6 | NP_195614.2 | 2 - 3 | 807 | 72 | |
GenBank mRNA | AF416569.1 | - | 2 - 3 | 504 | 72 | |
AK175482.1 | - | 2 - 3 | 577 | 72 | ||
AK176074.1 | - | 2 - 3 | 576 | 72 | ||
AK176272.1 | - | 2 - 3 | 575 | 72 | ||
AK176334.1 | - | 2 - 3 | 578 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |