Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120063.6 | NP_195614.2 | 4 - 5 | 980 | 83 | |
GenBank mRNA | AF416569.1 | - | 4 - 5 | 677 | 83 | |
AK175482.1 | - | 4 - 5 | 750 | 83 | ||
AK176074.1 | - | 4 - 5 | 749 | 83 | ||
AK176272.1 | - | 4 - 5 | 748 | 83 | ||
AK176334.1 | - | 4 - 5 | 751 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |