Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CESA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120095.4 | NP_195645.1 | 9 - 10 | 2531 | 124 | |
GenBank mRNA | AF027173.2 | - | 9 - 10 | 2011 | 124 | |
KJ139038.1 | - | 9 - 10 | 1841 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |