Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204036.1 | NP_001190965.1 | 8 - 9 | 1427 | 71 | |
NM_001342538.1 | NP_001320167.1 | 7 - 8 | 1405 | 71 | ||
NM_120097.3 | NP_568058.1 | 8 - 9 | 1325 | 71 | ||
GenBank mRNA | AF302675.1 | - | 8 - 9 | 1155 | 71 | |
AF370208.1 | - | 8 - 9 | 1290 | 71 | ||
AY133770.1 | - | 8 - 9 | 1137 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |