Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161045.2 | NP_001154517.1 | 18 - 19 | 1868 | 83 | |
NM_001202597.1 | NP_001189526.1 | 17 - 18 | 1799 | 83 | ||
NM_001335434.1 | NP_001324682.1 | 11 - 12 | 1280 | 83 | ||
NM_127032.2 | NP_179075.1 | 16 - 17 | 1929 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |