Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIC21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127089.4 | NP_565372.1 | 2 - 3 | 618 | 70 | |
GenBank mRNA | AY057510.1 | - | 2 - 3 | 598 | 70 | |
AY065215.1 | - | 2 - 3 | 598 | 70 | ||
AY122931.1 | - | 2 - 3 | 574 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |