Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFB1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127091.3 | NP_179133.1 | 1 - 2 | 427 | 117 | |
GenBank mRNA | AY054470.1 | - | 1 - 2 | 298 | 117 | |
AY093273.1 | - | 1 - 2 | 143 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |