Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127123.3 | NP_179164.1 | 3 - 4 | 1181 | 82 | |
GenBank mRNA | AB006032.1 | - | 3 - 4 | 1035 | 82 | |
AF360320.1 | - | 3 - 4 | 1048 | 82 | ||
AY045608.1 | - | 3 - 4 | 1046 | 82 | ||
AY093995.1 | - | 3 - 4 | 1024 | 82 | ||
AY142685.1 | - | 3 - 4 | 1024 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Probe spans exons |