Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127196.4 | NP_179235.1 | 5 - 6 | 1174 | 73 | |
NM_179629.2 | NP_849960.1 | 4 - 5 | 1009 | 73 | ||
GenBank mRNA | AF492662.1 | - | 5 - 6 | 867 | 73 | |
AY093236.1 | - | 5 - 6 | 915 | 73 | ||
BT008761.1 | - | 5 - 6 | 867 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |