Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP2;8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127238.3 | NP_179277.1 | 2 - 3 | 891 | 64 | |
GenBank mRNA | AK228097.1 | - | 2 - 3 | 644 | 64 | |
BT005214.1 | - | 2 - 3 | 578 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |