Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127244.4 | NP_179283.2 | 1 - 2 | 104 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |