Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036282.2 | NP_001031359.1 | 15 - 16 | 1694 | 60 | |
NM_127248.4 | NP_179287.3 | 15 - 16 | 1632 | 60 | ||
GenBank mRNA | BT002041.1 | - | 15 - 16 | 1620 | 60 | |
BT008804.1 | - | 15 - 16 | 1528 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |