Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036282.2 | NP_001031359.1 | 5 - 6 | 889 | 65 | |
NM_127248.4 | NP_179287.3 | 5 - 6 | 827 | 65 | ||
GenBank mRNA | BT002041.1 | - | 5 - 6 | 815 | 65 | |
BT008804.1 | - | 5 - 6 | 723 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |