Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335544.1 | NP_001318239.1 | 2 - 3 | 2294 | 87 | |
GenBank mRNA | AY495448.1 | - | 2 - 3 | 2269 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |