Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335545.1 | NP_001324056.1 | 7 - 8 | 2112 | 131 | |
NM_127284.3 | NP_565411.2 | 7 - 8 | 1913 | 131 | ||
GenBank mRNA | AY140007.1 | - | 7 - 8 | 1788 | 131 | |
BT002600.1 | - | 6 - 7 | 1525 | 131 | ||
U31835.1 | - | 6 - 7 | 1587 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |