Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127294.5 | NP_179331.1 | 4 - 5 | 557 | 71 | |
GenBank mRNA | BT024730.1 | - | 3 - 4 | 396 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |