Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATSERAT3;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127318.5 | NP_565421.1 | 1 - 2 | 404 | 153 | |
GenBank mRNA | AF112303.1 | - | 1 - 2 | 158 | 153 | |
AK176573.1 | - | 1 - 2 | 151 | 153 | ||
BT024717.1 | - | 1 - 2 | 135 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |