Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335556.1 | NP_001324196.1 | 7 - 8 | 1376 | 82 | |
NM_001335557.1 | NP_001324198.1 | 7 - 8 | 1485 | 82 | ||
NM_001335558.1 | NP_001324199.1 | 6 - 7 | 1740 | 82 | ||
NM_001335559.1 | NP_001324200.1 | 7 - 8 | 1663 | 82 | ||
NM_001335560.1 | NP_001324197.1 | 6 - 7 | 1697 | 82 | ||
NM_127332.3 | NP_179369.2 | 8 - 9 | 1448 | 82 | ||
GenBank mRNA | AB196961.1 | - | 6 - 7 | 1199 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |