Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336987.1 | NP_001324501.1 | 4 - 5 | 1277 | 114 | |
NM_180041.5 | NP_850372.4 | 5 - 6 | 1259 | 114 | ||
NM_180042.5 | NP_850373.4 | 5 - 6 | 967 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |