Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336987.1 | NP_001324501.1 | 5 - 6 | 1379 | 93 | |
NM_180041.5 | NP_850372.4 | 6 - 7 | 1361 | 93 | ||
NM_180042.5 | NP_850373.4 | 6 - 7 | 1069 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |