Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OEP37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129957.4 | NP_566003.1 | 3 - 4 | 829 | 76 | |
NM_180074.3 | NP_850405.2 | 3 - 4 | 772 | 76 | ||
GenBank mRNA | AY045937.1 | - | 3 - 4 | 710 | 76 | |
AY079409.1 | - | 3 - 4 | 687 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |