Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180076.2 | NP_850407.1 | 2 - 3 | 201 | 100 | |
GenBank mRNA | AY070449.1 | - | 2 - 3 | 193 | 100 | |
AY096566.1 | - | 2 - 3 | 161 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |