Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX11D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036467.3 | NP_001031544.1 | 5 - 6 | 545 | 96 | |
NM_130137.6 | NP_566055.1 | 4 - 5 | 614 | 96 | ||
NM_180110.5 | NP_850441.1 | 5 - 6 | 701 | 96 | ||
GenBank mRNA | AY054520.1 | - | 5 - 6 | 458 | 96 | |
AY084943.1 | - | 4 - 5 | 556 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |