Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180116.2 | NP_850447.1 | 10 - 11 | 1593 | 134 | |
GenBank mRNA | AY069912.1 | - | 10 - 11 | 1534 | 134 | |
BT001025.1 | - | 10 - 11 | 1444 | 134 | ||
EU249331.1 | - | 10 - 11 | 1444 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |