Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180116.2 | NP_850447.1 | 9 - 10 | 1304 | 91 | |
GenBank mRNA | AY069912.1 | - | 9 - 10 | 1245 | 91 | |
BT001025.1 | - | 9 - 10 | 1155 | 91 | ||
EU249331.1 | - | 9 - 10 | 1155 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |