Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337221.1 | NP_001318437.1 | 5 - 6 | 1013 | 80 | |
NM_180129.4 | NP_850460.1 | 5 - 6 | 739 | 80 | ||
NM_180130.2 | NP_850461.1 | 2 - 3 | 267 | 80 | ||
GenBank mRNA | AY519511.1 | - | 4 - 5 | 409 | 80 | |
BT001096.1 | - | 1 - 2 | 163 | 80 | ||
BT001105.1 | - | 1 - 2 | 163 | 80 | ||
U28422.1 | - | 5 - 6 | 646 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |