Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337223.1 | NP_001325392.1 | 3 - 4 | 2936 | 106 | |
NM_001337224.1 | NP_001325391.1 | 3 - 4 | 2877 | 106 | ||
NM_180132.3 | NP_850463.1 | 3 - 4 | 2942 | 106 | ||
NM_180133.2 | NP_850464.1 | 2 - 3 | 3042 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |