Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SWI3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337268.1 | NP_001324817.1 | 2 - 3 | 677 | 92 | |
NM_180145.3 | NP_850476.1 | 2 - 3 | 669 | 92 | ||
GenBank mRNA | AY062794.1 | - | 2 - 3 | 648 | 92 | |
AY081570.1 | - | 2 - 3 | 582 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |