Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036481.1 | NP_001031558.1 | 4 - 5 | 716 | 68 | |
NM_001202846.1 | NP_001189775.1 | 3 - 4 | 1031 | 68 | ||
NM_180150.2 | NP_850481.1 | 3 - 4 | 1121 | 68 | ||
GenBank mRNA | AK318961.1 | - | 4 - 5 | 720 | 68 | |
AY045774.1 | - | 3 - 4 | 562 | 68 | ||
AY074273.1 | - | 3 - 4 | 1016 | 68 | ||
AY096604.1 | - | 3 - 4 | 562 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |