Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337562.1 | NP_001325515.1 | 7 - 8 | 1085 | 86 | |
NM_111381.3 | NP_566246.1 | 7 - 8 | 1202 | 86 | ||
NM_180185.3 | NP_850516.1 | 7 - 8 | 1200 | 86 | ||
GenBank mRNA | AK226843.1 | - | 7 - 8 | 1160 | 86 | |
AY056292.1 | - | 7 - 8 | 1110 | 86 | ||
AY127022.1 | - | 7 - 8 | 1153 | 86 | ||
AY142674.1 | - | 6 - 7 | 801 | 86 | ||
BT000631.1 | - | 6 - 7 | 801 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |