Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111445.5 | NP_566256.1 | 7 - 8 | 1202 | 63 | |
NM_180188.3 | NP_850519.1 | 7 - 8 | 1148 | 63 | ||
GenBank mRNA | AJ245408.1 | - | 7 - 8 | 1124 | 63 | |
AY099723.1 | - | 7 - 8 | 1148 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |