Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180201.3 | NP_850532.1 | 4 - 5 | 608 | 105 | |
NM_180202.1 | NP_850533.1 | 4 - 5 | 410 | 105 | ||
GenBank mRNA | AY065244.1 | - | 4 - 5 | 407 | 105 | |
AY142587.1 | - | 4 - 5 | 385 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |