Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180201.3 | NP_850532.1 | 5 - 6 | 713 | 85 | |
NM_180202.1 | NP_850533.1 | 5 - 6 | 515 | 85 | ||
GenBank mRNA | AY065244.1 | - | 5 - 6 | 512 | 85 | |
AY142587.1 | - | 5 - 6 | 490 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |