Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111999.4 | NP_187773.1 | 2 - 3 | 1109 | 137 | |
NM_180230.1 | NP_850561.1 | 2 - 3 | 784 | 137 | ||
GenBank mRNA | AF149841.1 | - | 2 - 3 | 794 | 137 | |
AX765832.1 | - | 2 - 3 | 733 | 137 | ||
AY075649.1 | - | 2 - 3 | 784 | 137 | ||
AY101521.1 | - | 2 - 3 | 733 | 137 | ||
KJ138827.1 | - | 2 - 3 | 733 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |